missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Dibromo-4-nitroaniline 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 207°C |
| Color | Yellow |
| Quantity | 25 g |
| Formula Weight | 295.92 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,6-Dibromo-4-nitroaniline |
Safety and Handling
| EINECSNumber | (3)-0424 |
| RTECSNumber | BX2599000 |
| TSCA | Yes |