missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4-isopropylaniline, 98%

Catalog No. p-7022936
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1593206 5 g
AAA1593214 25 g
AAA1593222 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1593206 Supplier Thermo Scientific Chemicals Supplier No. A1593206

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 10546-65-3 | C9H11Br2N | 293.002 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 10546-65-3
Molecular Formula C9H11Br2N
Molecular Weight (g/mol) 293.002
MDL Number MFCD00015475
InChI Key CJEBZUFROMNDEK-UHFFFAOYSA-N
PubChem CID 139184
IUPAC Name 2,6-dibromo-4-propan-2-ylaniline
SMILES CC(C)C1=CC(=C(C(=C1)Br)N)Br

Specifications

Melting Point 45°C to 47°C
Quantity 5 g
Synonym 2,6-dibromo-4-isopropylaniline, 2,6-dibromo-4-methylethyl phenylamine, benzenamine,2,6-dibromo-4-1-methylethyl, benzenamine, 2,6-dibromo-4-1-methylethyl, 2,6-dibromocumidine, 2,6-dibromo-cumidine, acmc-20ao9h, 2,6-dibromo-4-iso-propylaniline, 2,6-dibromo-4-propan-2-yl aniline, 2,6-dibromo-4-1-methylethyl aniline
Formula Weight 293.01
Percent Purity 98%
Chemical Name or Material 2,6-Dibromo-4-isopropylaniline
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only