missing translation for 'onlineSavingsMsg'
Learn More

2,6-Dibromo-4,8-bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b']dithiophene 97.0+%, TCI America™

Catalog No. D4524200MG
Change view
Click to view available options
Quantity:
200 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D4524200MG 200 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D4524200MG Supplier TCI America Supplier No. D4524200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 1226782-13-3
Molecular Formula C26H36Br2O2S2
Molecular Weight (g/mol) 604.5
MDL Number MFCD17018552
InChI Key HMHJSVWOWIHEIP-UHFFFAOYSA-N
PubChem CID 51341874
IUPAC Name 2,6-dibromo-4,8-bis(2-ethylhexoxy)thieno[2,3-f][1]benzothiole
SMILES CCCCC(CC)COC1=C2C=C(SC2=C(C3=C1SC(=C3)Br)OCC(CC)CCCC)Br

Specifications

Quantity 200 mg
Formula Weight 604.50
Percent Purity ≥97.0% (HPLC)
Physical Form Liquid
Chemical Name or Material 2,6-Dibromo-4,8-bis(2-ethylhexyloxy)benzo[1,2-b:4,5-b′]dithiophene