accessibility menu, dialog, popup

UserName

H3CCH3OSSBrOCH3CH3Br

CAS RN 1226782-13-3

IUPAC Name: 5,11-dibromo-2,8-bis[(2-ethylhexyl)oxy]-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Synonyms: 5,11-dibromo-2,8-bis[(2-ethylhexyl)oxy]-4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Molecular Weight (g/mol): 604.5
Molecular Formula: C26H36Br2O2S2
InChi Key: HMHJSVWOWIHEIP-UHFFFAOYNA-N
SMILES: CCCCC(CC)COC1=C2SC(Br)=CC2=C(OCC(CC)CCCC)C2=C1C=C(Br)S2

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results