missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Diaminopurine Monohydrate 97.0+%, TCI America™
$46.60 - $119.30
Chemical Identifiers
| CAS | 402846-48-4 |
|---|---|
| Molecular Formula | C5H8N6O |
| Molecular Weight (g/mol) | 168.16 |
| MDL Number | MFCD00213668 |
| InChI Key | UKADKJJNEZZNGH-UHFFFAOYSA-N |
| PubChem CID | 19092948 |
| IUPAC Name | 7H-purine-2,6-diamine;hydrate |
| SMILES | C1=NC2=C(N1)C(=NC(=N2)N)N.O |
Chemical Identifiers
| 402846-48-4 | |
| 168.16 | |
| UKADKJJNEZZNGH-UHFFFAOYSA-N | |
| 7H-purine-2,6-diamine;hydrate |
| C5H8N6O | |
| MFCD00213668 | |
| 19092948 | |
| C1=NC2=C(N1)C(=NC(=N2)N)N.O |
Specifications
| 402846-48-4 | |
| C5H8N6O | |
| 5 g | |
| C1=NC2=C(N1)C(=NC(=N2)N)N.O | |
| 168.16 | |
| 150.15 | |
| Crystalline Powder |
| Yellow | |
| MFCD00213668 | |
| UKADKJJNEZZNGH-UHFFFAOYSA-N | |
| 7H-purine-2,6-diamine;hydrate | |
| 19092948 | |
| ≥97.0% (HPLC,T) | |
| 2,6-Diaminopurine Monohydrate |
Safety and Handling
RTECSNumber : UO7524000
TSCA : Yes