missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diaminoheptanedioic acid, 95%

Catalog No. p-3778136
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC235540010 1 g Glass bottle
AC235540050 5 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC235540010 Supplier Thermo Scientific Chemicals Supplier No. 235540010
Only null left

CAS: 583-93-7 | C7H14N2O4 | 190.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 583-93-7
Molecular Formula C7H14N2O4
Molecular Weight (g/mol) 190.2
MDL Number MFCD00002637
InChI Key GMKMEZVLHJARHF-UHFFFAOYSA-N
Synonym 2,6-diaminopimelic acid, heptanedioic acid, 2,6-diamino, diaminopimelic acid, dl-2,6-diaminoheptanedioic acid, m-dap, dl-alpha,epsilon-diaminopimelic acid, 1,5-diaminoheptanedioic acid, ll-diaminopimelate, d,l-diaminopimelate, ll-a2pm
PubChem CID 865
ChEBI CHEBI:23673
IUPAC Name 2,6-diaminoheptanedioic acid
SMILES C(CC(C(=O)O)N)CC(C(=O)O)N

Specifications

CAS Min % 95.0
CAS Max % 100.0
Melting Point 300°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 95%
Packaging Glass bottle
Linear Formula HOOCCH(NH2)3(CH2)3CH(NH2)COOH
Quantity 1 g
Solubility Information Solubility in water: 0.92% (21°C). Other solubilities: soluble in diluted acids and alkalies
Formula Weight 190.2
Percent Purity 95%
Physical Form Chunks and/or Powder
Chemical Name or Material 2, 6-Diaminoheptanedioic acid
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
EINECSNumber 209-524-6

RUO – Research Use Only