missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Diacetylpyridine 98.0+%, TCI America™
Supplier: TCI America D237725G
Specifications
| 2,6-Diacetylpyridine | |
| 81°C | |
| C9H9NO2 | |
| 25 g | |
| BEZVGIHGZPLGBL-UHFFFAOYSA-N | |
| 1-(6-acetylpyridin-2-yl)ethan-1-one | |
| 70790 | |
| ≥98.0% (GC) |
| 1129-30-2 | |
| 126°C | |
| MFCD00006304 | |
| 2,6-diacetylpyridine, 1,1'-pyridine-2,6-diyl diethanone, 1-6-acetylpyridin-2-yl ethanone, pyridine-2,6-diacetyl, 2,6-diacetyl pyridine, 1-6-acetyl-pyridin-2-yl-ethanone, ethanone, 1,1'-2,6-pyridinediyl bis, 1-6-acetylpyridin-2-yl ethan-1-one, 2,6diacetylpyridine, pubchem3144 | |
| CC(=O)C1=CC=CC(=N1)C(C)=O | |
| 163.18 | |
| 163.18 | |
| Crystalline Powder |
Chemical Identifiers
| 1129-30-2 | |
| 163.18 | |
| BEZVGIHGZPLGBL-UHFFFAOYSA-N | |
| 70790 | |
| CC(=O)C1=CC=CC(=N1)C(C)=O |
| C9H9NO2 | |
| MFCD00006304 | |
| 2,6-diacetylpyridine, 1,1'-pyridine-2,6-diyl diethanone, 1-6-acetylpyridin-2-yl ethanone, pyridine-2,6-diacetyl, 2,6-diacetyl pyridine, 1-6-acetyl-pyridin-2-yl-ethanone, ethanone, 1,1'-2,6-pyridinediyl bis, 1-6-acetylpyridin-2-yl ethan-1-one, 2,6diacetylpyridine, pubchem3144 | |
| 1-(6-acetylpyridin-2-yl)ethan-1-one |
Safety and Handling
TSCA : No