missing translation for 'onlineSavingsMsg'
Learn More

2,6-Diacetylpyridine 98.0+%, TCI America™

Catalog No. p-7127265
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D23775G 5 g
D237725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D23775G Supplier TCI America Supplier No. D23775G
Only null left

Chemical Identifiers

CAS 1129-30-2
Molecular Formula C9H9NO2
Molecular Weight (g/mol) 163.18
MDL Number MFCD00006304
InChI Key BEZVGIHGZPLGBL-UHFFFAOYSA-N
Synonym 2,6-diacetylpyridine, 1,1'-pyridine-2,6-diyl diethanone, 1-6-acetylpyridin-2-yl ethanone, pyridine-2,6-diacetyl, 2,6-diacetyl pyridine, 1-6-acetyl-pyridin-2-yl-ethanone, ethanone, 1,1'-2,6-pyridinediyl bis, 1-6-acetylpyridin-2-yl ethan-1-one, 2,6diacetylpyridine, pubchem3144
PubChem CID 70790
IUPAC Name 1-(6-acetylpyridin-2-yl)ethan-1-one
SMILES CC(=O)C1=CC=CC(=N1)C(C)=O

Specifications

Melting Point 81°C
Boiling Point 126°C
Quantity 5 g
Formula Weight 163.18
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Diacetylpyridine