missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butylphenol 98.0+%, TCI America™

Catalog No. p-7126513
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D094125G 25 g
D0941500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D094125G Supplier TCI America Supplier No. D094125G
Only null left

Chemical Identifiers

CAS 128-39-2
Molecular Formula C14H22O
Molecular Weight (g/mol) 206.33
MDL Number MFCD00008820
InChI Key DKCPKDPYUFEZCP-UHFFFAOYSA-N
Synonym 2,6-di-tert-butylphenol, 2,6-di-t-butylphenol, 2,6-bis tert-butyl phenol, 2,6-bis 1,1-dimethylethyl phenol, phenol, 2,6-bis 1,1-dimethylethyl, 2,6 di-tert-butylphenol, ethanox 701, isonox 103, ethyl 701, ethyl an 701
PubChem CID 31405
IUPAC Name 2,6-di-tert-butylphenol
SMILES CC(C)(C)C1=CC=CC(=C1O)C(C)(C)C

Specifications

Color White-Yellow
Boiling Point 253°C
Quantity 25 g
Formula Weight 206.33
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 2,6-Di-tert-butylphenol
EINECSNumber (3)-0521&(3)-0526
RTECSNumber SK8265000
TSCA Yes