missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Di-tert-butylphenol, 99%
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| CAS Min % | 98.5 |
| CAS Max % | 100.0 |
| Melting Point | 35°C to 38°C |
| Color | White to Yellow |
| Boiling Point | 253°C |
| Flash Point | 118°C |
| Infrared Spectrum | Authentic |
| Assay Percent Range | 99% |
| Packaging | Glass bottle |
| Linear Formula | [(CH3)3C]2C6H3OH |
| Show More |
Safety and Handling
| Hazard Category | GHS Signal Word: Warning |
| Hazard Statement | GHS H Statement Toxic to aquatic life with long lasting effects. Harmful if swallowed. |
| Precautionary Statement | GHS P Statement Avoid release to the environment. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. |
| EINECSNumber | 204-884- |
RUO – Research Use Only