missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methylpyrylium Trifluoromethanesulfonate 97.0+%, TCI America™

Catalog No. D24205G
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D24205G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D24205G Supplier TCI America Supplier No. D24205G
Only null left

Chemical Identifiers

CAS 59643-43-5
Molecular Formula C15H23F3O4S
Molecular Weight (g/mol) 356.4
MDL Number MFCD00229854
InChI Key OTVABNVGYCQZFO-UHFFFAOYSA-M
Synonym 2,6-di-tert-butyl-4-methylpyrylium trifluoromethanesulfonate, 2,6-ditert-butyl-4-methylpyrylium; trifluoromethanesulfonate, 2,6-di-tert-butyl-4-methyl-1??-pyran-1-ylium triflate, 2,6-di-tert-butyl-4-methylpyrylium triflate, 2,6-di-tert-butyl-4-methyl-1$l^ 4-pyran-1-ylium triflate, 2,6-di-tert-butyl-4-methylpyrylium trifluoromethane sulfonate, 2,6-di-tert-butyl-4-methylpyran-1-ium trifluoromethanesulfonate, 2,6-ditert-butyl-4-methyl-pyrylium; tris fluoranyl methanesulfonate, 2,6-di-tert-butyl-4-methyl-pyranylium, trifluoro-methanesulfonate
PubChem CID 2736809
IUPAC Name 2,6-ditert-butyl-4-methylpyrylium;trifluoromethanesulfonate
SMILES CC1=CC(=[O+]C(=C1)C(C)(C)C)C(C)(C)C.C(F)(F)(F)S(=O)(=O)[O-]

Specifications

Melting Point 172°C
Quantity 5 g
Formula Weight 356.40
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Di-tert-butyl-4-methylpyrylium Trifluoromethanesulfonate