accessibility menu, dialog, popup

UserName

CH3OH3CCH3H3CCH3H3CCH3FFFSOOO

CAS RN 59643-43-5

IUPAC Name: 2,6-di-tert-butyl-4-methyl-1λ⁴-pyran-1-ylium trifluoromethanesulfonate
Synonyms: 2,6-di-tert-butyl-4-methyl-1λ⁴-pyran-1-ylium triflate
Molecular Weight (g/mol): 356.4
Molecular Formula: C15H23F3O4S
InChi Key: OTVABNVGYCQZFO-UHFFFAOYSA-M
SMILES: [O-]S(=O)(=O)C(F)(F)F.CC1=CC(=[O+]C(=C1)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (1)
Physical Form 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results