Learn More
2,6-Di-tert-butyl-4-methylphenol, 99%
CAS: 128-37-0 | C15H24O | 220.356 g/mol
Supplier: Thermo Scientific Chemicals A1686330
Description
2,6-Di-tert-butyl-4-methylphenol is used as an antioxidant for cosmetics, vitamins, pharmaceuticals, rubber, oils and fats. It is used as a fuel additive in the petroleum industry and also used in hydraulic fluids, turbine and gear oils and jet fuels. It acts as a stabilizer in diethyl ether, tetrahydrofuran and other laboratory chemicals to prevent peroxide formation. It is effectively involved as a polymerization inhibitor in the process of oxidation of allyl alcohol to glycerine. It is utilized in the synthesis of organoaluminum compound, methylaluminum bis(2,6-di-tert-butyl-4-alkylphenoxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2,6-Di-tert-butyl-4-methylphenol is used as an antioxidant for cosmetics, vitamins, pharmaceuticals, rubber, oils and fats. It is used as a fuel additive in the petroleum industry and also used in hydraulic fluids, turbine and gear oils and jet fuels. It acts as a stabilizer in diethyl ether, tetrahydrofuran and other laboratory chemicals to prevent peroxide formation. It is effectively involved as a polymerization inhibitor in the process of oxidation of allyl alcohol to glycerine. It is utilized in the synthesis of organoaluminum compound, methylaluminum bis(2,6-di-tert-butyl-4-alkylphenoxide.
Solubility
Soluble in methanol, toluene, isopropanol, methyl ethyl ketone, acetone, benzene, petroleum ether and ethanol. Insoluble in water.
Notes
Light-sensitive. Keep the container tightly closed in a cool, dry and well-ventilated place. Incompatible with acid chlorides, acid anhydrides, oxidizing agents, bases, brass, copper and copper alloys.
Specifications
| 2,6-Di-tert-butyl-4-methylphenol | |
| 69°C to 72°C | |
| 264°C to 265°C | |
| Mild | |
| MFCD00011644 | |
| UN3077 | |
| 14,1548 | |
| Soluble in methanol,toluene,isopropanol,methyl ethyl ketone,acetone,benzene,petroleum ether and ethanol. Insoluble in water. | |
| CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C | |
| 220.356 | |
| CHEBI:34247 | |
| 99% |
| 128-37-0 | |
| 1.048 | |
| 127°C (260°F) | |
| C15H24O | |
| 250 g | |
| 1911640 | |
| 2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht | |
| NLZUEZXRPGMBCV-UHFFFAOYSA-N | |
| 2,6-ditert-butyl-4-methylphenol | |
| 31404 | |
| 220.36 |
Chemical Identifiers
| 128-37-0 | |
| 220.356 | |
| NLZUEZXRPGMBCV-UHFFFAOYSA-N | |
| 31404 | |
| 2,6-ditert-butyl-4-methylphenol |
| C15H24O | |
| MFCD00011644 | |
| 2,6-di-tert-butyl-4-methylphenol, butylated hydroxytoluene, butylhydroxytoluene, 2,6-di-tert-butyl-p-cresol, 2,6-di-t-butyl-4-methylphenol, ionol, dbpc, dibunol, stavox, bht | |
| CHEBI:34247 | |
| CC1=CC(=C(C(=C1)C(C)(C)C)O)C(C)(C)C |
Safety and Handling
GHS H Statement
H302-H319
Harmful if swallowed.
Causes serious eye irritation.
P260-P271-P304+P340-P312-P314-P501c
H335-H373
DOTInformation : Transport Hazard Class: 9; Packing Group: III; Proper Shipping Name: ENVIRONMENTALLY HAZARDOUS SUBSTANCES, SOLID, N.O.S.
EINECSNumber : 204-881-4
RTECSNumber : GO7875000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only