missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers) 95.0+%, TCI America™

Catalog No. d43851ml
Change view
Click to view available options
Quantity:
1 mL
5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D43851ML 1 mL
D43855ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D43851ML Supplier TCI America Supplier No. D43851ML
Only null left

Chemical Identifiers

CAS 23790-39-8
Molecular Formula C15H28O
Molecular Weight (g/mol) 224.388
InChI Key LDPDMUPXRDWOPE-UHFFFAOYSA-N
PubChem CID 228974
IUPAC Name 2,6-ditert-butyl-4-methylcyclohexan-1-one
SMILES CC1CC(C(=O)C(C1)C(C)(C)C)C(C)(C)C

Specifications

Color Colorless
Boiling Point 134°C
Quantity 1 mL
Formula Weight 224.39
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,6-Di-tert-butyl-4-methylcyclohexanone (mixture of isomers)