accessibility menu, dialog, popup

UserName

CH3OH3CCH3H3CCH3H3CCH3

CAS RN 23790-39-8

IUPAC Name: 2,6-di-tert-butyl-4-methylcyclohexan-1-one
Synonyms: 2,6-di-tert-butyl-4-methylcyclohexan-1-one
Molecular Weight (g/mol): 224.39
Molecular Formula: C15H28O
InChi Key: LDPDMUPXRDWOPE-UHFFFAOYNA-N
SMILES: CC1CC(C(=O)C(C1)C(C)(C)C)C(C)(C)C
Percent Purity 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results