missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methoxyphenol 98.0+%, TCI America™

Catalog No. D181125G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D181125G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. D181125G Supplier TCI America Supplier No. D181125G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 489-01-0
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.355
MDL Number MFCD00008824
InChI Key SLUKQUGVTITNSY-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methoxyphenol, 3,5-di-tert-butyl-4-hydroxyanisole, topanol 354, phenol, 2,6-bis 1,1-dimethylethyl-4-methoxy, 3,5-di-t-butyl-4-hydroxyanisole, unii-616072tmxy, ccris 5219, 2,6-bis 1,1-dimethylethyl-4-methoxyphenol, phenol, 2,6-di-tert-butyl-4-methoxy, chembl98253
PubChem CID 10269
IUPAC Name 2,6-ditert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)OC

Specifications

Melting Point 105°C
Boiling Point 140°C
Quantity 25 g
Formula Weight 236.36
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Di-tert-butyl-4-methoxyphenol [Oxidation inhibitor]