missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di-tert-butyl-4-methoxyphenol, 97%

Catalog No. AC294160250
Change view
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC294160250 25 g Glass bottle
AC294161000 100 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC294160250 Supplier Thermo Scientific Chemicals Supplier No. 294160250
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 489-01-0 | C15H24O2 | 236.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 489-01-0
Molecular Formula C15H24O2
Molecular Weight (g/mol) 236.35
MDL Number MFCD00008824
InChI Key SLUKQUGVTITNSY-UHFFFAOYSA-N
Synonym 2,6-di-tert-butyl-4-methoxyphenol, 3,5-di-tert-butyl-4-hydroxyanisole, topanol 354, phenol, 2,6-bis 1,1-dimethylethyl-4-methoxy, 3,5-di-t-butyl-4-hydroxyanisole, unii-616072tmxy, ccris 5219, 2,6-bis 1,1-dimethylethyl-4-methoxyphenol, phenol, 2,6-di-tert-butyl-4-methoxy, chembl98253
PubChem CID 10269
IUPAC Name 2,6-ditert-butyl-4-methoxyphenol
SMILES CC(C)(C)C1=CC(=CC(=C1O)C(C)(C)C)OC

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 103°C to 106°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 25 g
Fieser 16,110; 16,58
Solubility Information Solubility in water: practically insoluble
Formula Weight 236.35
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 2, 6-Di-tert-butyl-4-methoxyphenol, 97%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and
EINECSNumber 207-693-0
RTECSNumber SJ7785668
TSCA TSCA

RUO – Research Use Only