missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Di(1-pyrazolyl)pyridine 98.0+%, TCI America™
$265.67 - $910.61
Chemical Identifiers
| CAS | 123640-38-0 |
|---|---|
| Molecular Formula | C11H9N5 |
| Molecular Weight (g/mol) | 211.228 |
| InChI Key | OMNFEFLKDQUMRN-UHFFFAOYSA-N |
| PubChem CID | 11816656 |
| IUPAC Name | 2,6-di(pyrazol-1-yl)pyridine |
| SMILES | C1=CC(=NC(=C1)N2C=CC=N2)N3C=CC=N3 |
Chemical Identifiers
| 123640-38-0 | |
| 211.228 | |
| 11816656 | |
| C1=CC(=NC(=C1)N2C=CC=N2)N3C=CC=N3 |
| C11H9N5 | |
| OMNFEFLKDQUMRN-UHFFFAOYSA-N | |
| 2,6-di(pyrazol-1-yl)pyridine |
Specifications
| 123640-38-0 | |
| White | |
| 1 g | |
| C1=CC(=NC(=C1)N2C=CC=N2)N3C=CC=N3 | |
| 211.228 | |
| 211.23 | |
| Crystalline Powder |
| 139°C | |
| C11H9N5 | |
| OMNFEFLKDQUMRN-UHFFFAOYSA-N | |
| 2,6-di(pyrazol-1-yl)pyridine | |
| 11816656 | |
| ≥98.0% (GC) | |
| 2,6-Di(1-pyrazolyl)pyridine |
Safety and Handling
TSCA : No