accessibility menu, dialog, popup

UserName

NNNNN

CAS RN 123640-38-0

IUPAC Name: 2,6-bis(1H-pyrazol-1-yl)pyridine
Synonyms: 2,6-bis(pyrazol-1-yl)pyridine
Molecular Weight (g/mol): 211.23
Molecular Formula: C11H9N5
InChi Key: OMNFEFLKDQUMRN-UHFFFAOYSA-N
SMILES: C1=CN(N=C1)C1=CC=CC(=N1)N1C=CC=N1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results