missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Di(1-pyrazolyl)pyridine 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 139°C |
| Color | White |
| Quantity | 5 g |
| Formula Weight | 211.23 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | 2,6-Di(1-pyrazolyl)pyridine |
Safety and Handling
| TSCA | No |