missing translation for 'onlineSavingsMsg'
Learn More

2,6-Di(1-pyrazolyl)pyridine 98.0+%, TCI America™

Catalog No. D46721G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D46721G 1 g
D46725G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D46721G Supplier TCI America Supplier No. D46721G
Only null left

Chemical Identifiers

CAS 123640-38-0
Molecular Formula C11H9N5
Molecular Weight (g/mol) 211.228
InChI Key OMNFEFLKDQUMRN-UHFFFAOYSA-N
PubChem CID 11816656
IUPAC Name 2,6-di(pyrazol-1-yl)pyridine
SMILES C1=CC(=NC(=C1)N2C=CC=N2)N3C=CC=N3

Specifications

Melting Point 139°C
Color White
Quantity 1 g
Formula Weight 211.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Di(1-pyrazolyl)pyridine