missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)pyridine, 97%

Catalog No. AAL1944603
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1944603 1 g
AAL19446MD 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL1944603 Supplier Thermo Scientific Chemicals Supplier No. L1944603

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 455-00-5 | C7H3F6N | 215.098 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 455-00-5
Molecular Formula C7H3F6N
Molecular Weight (g/mol) 215.098
MDL Number MFCD00236675
InChI Key YPDVFTXBESQIPJ-UHFFFAOYSA-N
PubChem CID 589864
IUPAC Name 2,6-bis(trifluoromethyl)pyridine
SMILES C1=CC(=NC(=C1)C(F)(F)F)C(F)(F)F

Specifications

Melting Point 55°C to 58°C
Boiling Point 82°C (18 mmHg)
Flash Point 67°C (153°F)
Quantity 1 g
Beilstein 188806
Synonym 2,6-bis trifluoromethyl pyridine, 2,6-bis trifluoromethyl-pyridine, pubchem7696, acmc-209k2t, pyridine,2,6-bis trifluoromethyl, pyridine, 2,6-bis trifluoromethyl
Formula Weight 215.1
Percent Purity 97%
Chemical Name or Material 2,6-Bis(trifluoromethyl)pyridine
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H332-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only