missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(trifluoromethyl)benzoic acid, 98%

Catalog No. AAB2048606
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2048606 5 g
AAB2048603 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2048606 Supplier Thermo Scientific Chemicals Supplier No. B2048606
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 24821-22-5 | C9H4F6O2 | 258.119 g/mol

2,6-Bis(trifluoromethyl)benzoic acid is used as an organic intermediate in synthesis. It is also used as a pharmaceutical intermediate and organic intermediate in chemical synthesis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,6-Bis(trifluoromethyl)benzoic acid is used as an organic intermediate in synthesis. It is also used as a pharmaceutical intermediate and organic intermediate in chemical synthesis.

Solubility
Soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 24821-22-5
Molecular Formula C9H4F6O2
Molecular Weight (g/mol) 258.119
MDL Number MFCD00000376
InChI Key XZNLSDPNMNWCRE-UHFFFAOYSA-N
Synonym 2,6-bis trifluoromethyl benzoic acid, benzoic acid, 2,6-bis trifluoromethyl, 2,6-bis-trifluoromethylbenzoic acid, 2,6-di trifluoromethyl benzoic acid, 2,6-bis-trifluoromethyl-benzoic acid, 2,6-bis trifluoromethyl-benzoic acid, pubchem4988, acmc-1co7v, rarechem al bo 0513, timtec-bb sbb000962
PubChem CID 90618
IUPAC Name 2,6-bis(trifluoromethyl)benzoic acid
SMILES C1=CC(=C(C(=C1)C(F)(F)F)C(=O)O)C(F)(F)F

Specifications

Melting Point 136°C to 140°C
Odor Odorless
Quantity 5 g
Beilstein 2057459
Solubility Information Soluble in water.
Formula Weight 258.12
Percent Purity 98%
Chemical Name or Material 2,6-Bis(trifluoromethyl)benzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 246-479-1
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only