missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Bis(bromomethyl)pyridine 99.0+%, TCI America™
$121.16 - $425.00
Chemical Identifiers
| CAS | 7703-74-4 |
|---|---|
| Molecular Formula | C7H7Br2N |
| Molecular Weight (g/mol) | 264.948 |
| MDL Number | MFCD00191795 |
| InChI Key | QUTSYCOAZVHGGT-UHFFFAOYSA-N |
| PubChem CID | 603610 |
| IUPAC Name | 2,6-bis(bromomethyl)pyridine |
| SMILES | C1=CC(=NC(=C1)CBr)CBr |
Chemical Identifiers
| 7703-74-4 | |
| 264.948 | |
| QUTSYCOAZVHGGT-UHFFFAOYSA-N | |
| 2,6-bis(bromomethyl)pyridine |
| C7H7Br2N | |
| MFCD00191795 | |
| 603610 | |
| C1=CC(=NC(=C1)CBr)CBr |
Specifications
| 7703-74-4 | |
| White-Yellow | |
| MFCD00191795 | |
| QUTSYCOAZVHGGT-UHFFFAOYSA-N | |
| 2,6-bis(bromomethyl)pyridine | |
| 603610 | |
| ≥99.0% (GC,T) | |
| 2,6-Bis(bromomethyl)pyridine |
| 87°C | |
| C7H7Br2N | |
| 1 g | |
| C1=CC(=NC(=C1)CBr)CBr | |
| 264.948 | |
| 264.95 | |
| Crystalline Powder |
Safety and Handling
TSCA : No