missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(bromomethyl)pyridine 99.0+%, TCI America™

Catalog No. B19361G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B19361G 1 g
B19365G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B19361G Supplier TCI America Supplier No. B19361G
Only null left

Chemical Identifiers

CAS 7703-74-4
Molecular Formula C7H7Br2N
Molecular Weight (g/mol) 264.948
MDL Number MFCD00191795
InChI Key QUTSYCOAZVHGGT-UHFFFAOYSA-N
PubChem CID 603610
IUPAC Name 2,6-bis(bromomethyl)pyridine
SMILES C1=CC(=NC(=C1)CBr)CBr

Specifications

Melting Point 87°C
Color White-Yellow
Quantity 1 g
Formula Weight 264.95
Percent Purity ≥99.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(bromomethyl)pyridine