missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis(4-azidobenzylidene)cyclohexanone 85.0+%, TCI America™

Catalog No. B111025G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B111025G 25 g
B11105G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B111025G Supplier TCI America Supplier No. B111025G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

(wetted with ca. 30% Water)(unit weight on dry weight basis) [Research for Photosensitive Material]

Chemical Identifiers

CAS 20237-98-3
Molecular Formula C20H16N6O
Molecular Weight (g/mol) 356.39
MDL Number MFCD00059878
InChI Key UZNOMHUYXSAUPB-UNZYHPAISA-N
Synonym 2,6-Di(4-azidobenzal)cyclohexanone
PubChem CID 22788445
IUPAC Name (2E,6E)-2,6-bis[(4-azidophenyl)methylidene]cyclohexan-1-one
SMILES [N-]=[N+]=NC1=CC=C(\C=C2/CCC\C(=C/C3=CC=C(C=C3)N=[N+]=[N-])C2=O)C=C1

Specifications

Quantity 25 g
UN Number 1325
Formula Weight 356.39
Percent Purity ≥85.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis(4-azidobenzylidene)cyclohexanone (wetted with ca. 30% Water) (unit weight on dry weight basi
EINECSNumber (4)-0193
TSCA Yes