accessibility menu, dialog, popup

UserName

CAS RN 20237-98-3

IUPAC Name: (2E,6E)-2,6-bis[(4-azidophenyl)methylidene]cyclohexan-1-one
Synonyms: (2E,6E)-2,6-bis[(4-azidophenyl)methylidene]cyclohexan-1-one
Molecular Weight (g/mol): 356.39
Molecular Formula: C20H16N6O
InChi Key: UZNOMHUYXSAUPB-UNZYHPAISA-N
SMILES: [N-]=[N+]=NC1=CC=C(\C=C2/CCC\C(=C/C3=CC=C(C=C3)N=[N+]=[N-])C2=O)C=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results