missing translation for 'onlineSavingsMsg'
Learn More

2,6-Bis[(2S,5S)-4,4-diphenyl-1-aza-3-oxabicyclo[3.3.0]octan-2-yl]pyridine 90.0+%, TCI America™

Catalog No. B422850MG
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B422850MG 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B422850MG Supplier TCI America Supplier No. B422850MG
Only null left

Chemical Identifiers

CAS 1450841-27-6
Molecular Formula C41H39N3O2
Molecular Weight (g/mol) 605.78
MDL Number MFCD27976812
InChI Key UFOJHPHCTXHMFQ-UHFFFAOYNA-N
PubChem CID 91972044
IUPAC Name 2,6-bis({1,1-diphenyl-hexahydropyrrolo[1,2-c][1,3]oxazol-3-yl})pyridine
SMILES C1CC2N(C1)C(OC2(C1=CC=CC=C1)C1=CC=CC=C1)C1=CC=CC(=N1)C1OC(C2CCCN12)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Color White-Yellow
Quantity 50 mg
Formula Weight 605.78
Percent Purity ≥90.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,6-Bis[(2S,5S)-4,4-diphenyl-1-aza-3-oxabicyclo[3.3.0]octan-2-yl]pyridine