missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,6-Bis(2-benzimidazolyl)pyridine 98.0+%, TCI America™
$208.78 - $831.37
Chemical Identifiers
| CAS | 28020-73-7 |
|---|---|
| Molecular Formula | C19H13N5 |
| Molecular Weight (g/mol) | 311.35 |
| MDL Number | MFCD00134500 |
| InChI Key | JBKICBDXAZNSKA-UHFFFAOYSA-N |
| PubChem CID | 738986 |
| IUPAC Name | 2-[6-(1H-1,3-benzodiazol-2-yl)pyridin-2-yl]-1H-1,3-benzodiazole |
| SMILES | N1C2=CC=CC=C2N=C1C1=CC=CC(=N1)C1=NC2=CC=CC=C2N1 |
Chemical Identifiers
| 28020-73-7 | |
| 311.35 | |
| JBKICBDXAZNSKA-UHFFFAOYSA-N | |
| 2-[6-(1H-1,3-benzodiazol-2-yl)pyridin-2-yl]-1H-1,3-benzodiazole |
| C19H13N5 | |
| MFCD00134500 | |
| 738986 | |
| N1C2=CC=CC=C2N=C1C1=CC=CC(=N1)C1=NC2=CC=CC=C2N1 |
Specifications
| 28020-73-7 | |
| White-Yellow | |
| MFCD00134500 | |
| 2,2′C-(2,6-Pyridinediyl)bis(benzimidazole) | |
| N1C2=CC=CC=C2N=C1C1=CC=CC(=N1)C1=NC2=CC=CC=C2N1 | |
| 311.35 | |
| 311.35 | |
| Crystalline Powder |
| 353°C | |
| C19H13N5 | |
| 1 g | |
| JBKICBDXAZNSKA-UHFFFAOYSA-N | |
| 2-[6-(1H-1,3-benzodiazol-2-yl)pyridin-2-yl]-1H-1,3-benzodiazole | |
| 738986 | |
| ≥98.0% (HPLC) | |
| 2,6-Bis(2-benzimidazolyl)pyridine |
Safety and Handling
TSCA : No