missing translation for 'onlineSavingsMsg'
Learn More

2(5H)-Furanone, 96%

Catalog No. p-7046106
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL1314203 1 g
AAL1314206 5 g
AAL1314214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL1314203 Supplier Thermo Scientific Chemicals Supplier No. L1314203

CAS: 497-23-4 | C4H4O2 | 84.07 g/mol

2(5H)-Furanone is employed in the preparation of 5-substituted 2(5H) furanones (gamma-butenolides) catalyzed by bifunctional aminothiourea and aminosquaramide organocatalysts through direct aldol reaction with aromatic aldehydes. Further, it is used in the synthesis of lignans by Michael addition reactions. In addition to this, it is used in three-component Michael-Aldol reactions with an aldehyde and thiolate or carbanion.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2(5H)-Furanone is employed in the preparation of 5-substituted 2(5H) furanones (gamma-butenolides) catalyzed by bifunctional aminothiourea and aminosquaramide organocatalysts through direct aldol reaction with aromatic aldehydes. Further, it is used in the synthesis of lignans by Michael addition reactions. In addition to this, it is used in three-component Michael-Aldol reactions with an aldehyde and thiolate or carbanion.

Solubility
Immiscible with water.

Notes
Light and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 497-23-4
Molecular Formula C4H4O2
Molecular Weight (g/mol) 84.07
MDL Number MFCD00005376
InChI Key VIHAEDVKXSOUAT-UHFFFAOYSA-N
Synonym 2 5h-furanone, furan-2 5h-one, butenolide, 2-butenolide, gamma-crotonolactone, 2-buten-4-olide, isocrotonolactone, 2-oxo-2,5-dihydrofuran, crotonolactone, 2-5h-furanone
PubChem CID 10341
ChEBI CHEBI:38118
IUPAC Name 2H-furan-5-one
SMILES O=C1OCC=C1

Specifications

Melting Point 3°C to 4°C
Density 1.197
Boiling Point 212°C to 214°C
Flash Point 101°C (213°F)
Refractive Index 1.469
Quantity 1 g
Beilstein 383585
Sensitivity Light sensitive
Solubility Information Immiscible with water.
Formula Weight 84.07
Percent Purity 98%
Chemical Name or Material 2(5H)-Furanone
Show More Show Less
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 207-839-3
RTECSNumber LU3453000
TSCA No
Recommended Storage Keep cold; Store under Nitrogen

RUO – Research Use Only