missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole 98.0+%, TCI America™
$96.94 - $781.03
Chemical Identifiers
| CAS | 3147-76-0 |
|---|---|
| Molecular Formula | C16H17N3O |
| Molecular Weight (g/mol) | 267.332 |
| MDL Number | MFCD03844795 |
| InChI Key | WXHVQMGINBSVAY-UHFFFAOYSA-N |
| PubChem CID | 76605 |
| IUPAC Name | 2-(benzotriazol-2-yl)-4-tert-butylphenol |
| SMILES | CC(C)(C)C1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2 |
Chemical Identifiers
| 3147-76-0 | |
| 267.332 | |
| WXHVQMGINBSVAY-UHFFFAOYSA-N | |
| 2-(benzotriazol-2-yl)-4-tert-butylphenol |
| C16H17N3O | |
| MFCD03844795 | |
| 76605 | |
| CC(C)(C)C1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2 |
Specifications
| 3147-76-0 | |
| White-Yellow | |
| MFCD03844795 | |
| WXHVQMGINBSVAY-UHFFFAOYSA-N | |
| 2-(benzotriazol-2-yl)-4-tert-butylphenol | |
| 76605 | |
| ≥98.0% (T) | |
| 2-(5-tert-Butyl-2-hydroxyphenyl)benzotriazole |
| 97°C | |
| C16H17N3O | |
| 25 g | |
| CC(C)(C)C1=CC(=C(C=C1)O)N2N=C3C=CC=CC3=N2 | |
| 267.332 | |
| 267.33 | |
| Crystalline Powder |
Safety and Handling
EINECSNumber : (5)-3606
TSCA : Yes