missing translation for 'onlineSavingsMsg'
Learn More

2,5-Norbornadiene Palladium(II) Dichloride 97.0+%, TCI America™

Catalog No. N08421G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N08421G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N08421G Supplier TCI America Supplier No. N08421G
Only null left

Chemical Identifiers

CAS 12317-46-3
Molecular Formula C7H8Cl2Pd
Molecular Weight (g/mol) 269.461
MDL Number MFCD00134222
InChI Key BNCRZJHZZCMDNP-UHFFFAOYSA-L
Synonym Dichloro-2,5-norbornadiene Palladium(II), 2,5-Norbornadienedichloropalladium(II)
PubChem CID 11346242
IUPAC Name bicyclo[2.2.1]hepta-2,5-diene;dichloropalladium
SMILES C1C2C=CC1C=C2.Cl[Pd]Cl

Specifications

Melting Point 260°C
Color Yellow
Quantity 1 g
Formula Weight 269.46
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Norbornadiene Palladium(II) Dichloride