CAS RN 12317-46-3
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
269.46
Molecular Formula:
C7H8Cl2Pd
InChi Key:
BNCRZJHZZCMDNP-UHFFFAOYSA-L
SMILES:
[Cl-].[Cl-].[Pd++].[CH]1[CH]C2[CH][CH]C1C2
1
–
1
of
1
results
1
2,5-Norbornadiene Palladium(II) Dichloride 97.0+%, TCI America™
CAS: 12317-46-3 Molecular Formula: C7H8Cl2Pd Molecular Weight (g/mol): 269.461 MDL Number: MFCD00134222 InChI Key: BNCRZJHZZCMDNP-UHFFFAOYSA-L Synonym: Dichloro-2,5-norbornadiene Palladium(II), 2,5-Norbornadienedichloropalladium(II) PubChem CID: 11346242 IUPAC Name: bicyclo[2.2.1]hepta-2,5-diene;dichloropalladium SMILES: C1C2C=CC1C=C2.Cl[Pd]Cl