missing translation for 'onlineSavingsMsg'
Learn More

2,5-Hexanediol 98.0+%, TCI America™

Catalog No. p-7130339
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
H01005ML 5 mL
H010025ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. H01005ML Supplier TCI America Supplier No. H01005ML
Only null left

Chemical Identifiers

CAS 2935-44-6
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00004557
InChI Key OHMBHFSEKCCCBW-UHFFFAOYSA-N
Synonym 2,5-hexanediol, diisopropanol, 2,5-dihydroxyhexane, 2,5-hexyleneglycol, 2,5-hexanediol, mixture of isomers, 2,5-hexandiol, hexan-2,5-diol, 2,5-hexylene glycol, acmc-1acz0, acmc-209e5m
PubChem CID 18049
ChEBI CHEBI:84894
IUPAC Name hexane-2,5-diol
SMILES CC(CCC(C)O)O

Specifications

Color Yellow
Boiling Point 221°C
Quantity 5 mL
Formula Weight 118.18
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 2,5-Hexanediol
EINECSNumber (2)-0240
RTECSNumber MO2275000
TSCA Yes