accessibility menu, dialog, popup

UserName

H3COHCH3OH

CAS RN 2935-44-6

IUPAC Name: hexane-2,5-diol
Synonyms: 2,5-hexanediol
Molecular Weight (g/mol): 118.18
Molecular Formula: C6H14O2
InChi Key: OHMBHFSEKCCCBW-UHFFFAOYNA-N
SMILES: CC(O)CCC(C)O
Boiling Point 
  • (2)
  • (2)
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)