missing translation for 'onlineSavingsMsg'
Learn More

2,5-Hexanediol, mixture of isomers, 98%

Catalog No. p-7028253
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2547009 10 g
AAB2547018 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2547009 Supplier Thermo Scientific Chemicals Supplier No. B2547009

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 2935-44-6 | C6H14O2 | 118.176 g/mol

2,5-Hexanediol is used as an organic chemical synthesis intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Hexanediol is used as an organic chemical synthesis intermediate.

Solubility
Fully miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 2935-44-6
Molecular Formula C6H14O2
Molecular Weight (g/mol) 118.176
MDL Number MFCD00004557
InChI Key OHMBHFSEKCCCBW-UHFFFAOYSA-N
Synonym 2,5-hexanediol, diisopropanol, 2,5-dihydroxyhexane, 2,5-hexyleneglycol, 2,5-hexanediol, mixture of isomers, 2,5-hexandiol, hexan-2,5-diol, 2,5-hexylene glycol, acmc-1acz0, acmc-209e5m
PubChem CID 18049
ChEBI CHEBI:84894
IUPAC Name hexane-2,5-diol
SMILES CC(CCC(C)O)O

Specifications

Density 0.961
Boiling Point 216°C to 218°C
Flash Point 101°C (215°F)
Refractive Index 1.447
Linear Formula CH3CH(OH)CH2CH2CH(OH)CH3
Quantity 10 g
Solubility Information Fully miscible in water.
Formula Weight 118.18
Percent Purity 98%
Chemical Name or Material 2,5-Hexanediol, mixture of isomers
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 220-910-3
RTECSNumber MO2275000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only