missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dithiobiurea 95.0+%, TCI America™
$117.44 - $225.25
Chemical Identifiers
| CAS | 142-46-1 |
|---|---|
| Molecular Formula | C2H6N4S2 |
| Molecular Weight (g/mol) | 150.22 |
| MDL Number | MFCD00014471 |
| InChI Key | KCOYHFNCTWXETP-UHFFFAOYSA-N |
| Synonym | 2,5-dithiobiurea, dithiobiurea, bithiourea, dithiourea, 1,2-hydrazinedicarbothioamide, bis thiourea, hydrazodicarbothioamide, 2,5-dithiodiurea, hydrazine-1,2-bis carbothioamide, dithiocarbamoylhydrazine |
| PubChem CID | 2724564 |
| IUPAC Name | (carbamothioylamino)thiourea |
| SMILES | NC(=S)NNC(N)=S |
Chemical Identifiers
| 142-46-1 | |
| 150.22 | |
| KCOYHFNCTWXETP-UHFFFAOYSA-N | |
| 2724564 | |
| NC(=S)NNC(N)=S |
| C2H6N4S2 | |
| MFCD00014471 | |
| 2,5-dithiobiurea, dithiobiurea, bithiourea, dithiourea, 1,2-hydrazinedicarbothioamide, bis thiourea, hydrazodicarbothioamide, 2,5-dithiodiurea, hydrazine-1,2-bis carbothioamide, dithiocarbamoylhydrazine | |
| (carbamothioylamino)thiourea |
Specifications
| 142-46-1 | |
| White-Yellow | |
| MFCD00014471 | |
| 2,5-dithiobiurea, dithiobiurea, bithiourea, dithiourea, 1,2-hydrazinedicarbothioamide, bis thiourea, hydrazodicarbothioamide, 2,5-dithiodiurea, hydrazine-1,2-bis carbothioamide, dithiocarbamoylhydrazine | |
| NC(=S)NNC(N)=S | |
| 150.22 | |
| 150.22 | |
| Crystalline Powder |
| 196°C | |
| C2H6N4S2 | |
| 10 g | |
| KCOYHFNCTWXETP-UHFFFAOYSA-N | |
| (carbamothioylamino)thiourea | |
| 2724564 | |
| ≥95.0% (HPLC) | |
| 2,5-Dithiobiurea |
Safety and Handling
RTECSNumber : EC1460000
TSCA : Yes