accessibility menu, dialog, popup

UserName

H2NSNHHNNH2S

CAS RN 142-46-1

IUPAC Name: (carbamothioylamino)thiourea
Synonyms: 1,2-hydrazinecarbothioamide
Molecular Weight (g/mol): 150.22
Molecular Formula: C2H6N4S2
InChi Key: KCOYHFNCTWXETP-UHFFFAOYSA-N
SMILES: NC(=S)NNC(N)=S
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (2)