missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dithiobiurea 95.0+%, TCI America™

Catalog No. D130825G
Change view
Click to view available options
Quantity:
10 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D130825G 25 g
D130810G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D130825G Supplier TCI America Supplier No. D130825G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 142-46-1
Molecular Formula C2H6N4S2
Molecular Weight (g/mol) 150.22
MDL Number MFCD00014471
InChI Key KCOYHFNCTWXETP-UHFFFAOYSA-N
Synonym 2,5-dithiobiurea, dithiobiurea, bithiourea, dithiourea, 1,2-hydrazinedicarbothioamide, bis thiourea, hydrazodicarbothioamide, 2,5-dithiodiurea, hydrazine-1,2-bis carbothioamide, dithiocarbamoylhydrazine
PubChem CID 2724564
IUPAC Name (carbamothioylamino)thiourea
SMILES NC(=S)NNC(N)=S

Specifications

Melting Point 196°C
Color White-Yellow
Quantity 25 g
Formula Weight 150.22
Percent Purity ≥95.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dithiobiurea
RTECSNumber EC1460000
TSCA Yes