missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dithiobiurea, 97%

Catalog No. AAA1666714
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1666714 25 g
AAA1666722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1666714 Supplier Thermo Scientific Chemicals Supplier No. A1666714

CAS: 142-46-1 | C2H6N4S2 | 150.22 g/mol

2,5-Dithiobiurea is used in radiopharmaceutical imaging, inhibiting enzyme function, kidney function study and to treat toxic metal poisoning. It is also used in co-crystals development and in the field of nonlinear optics to generate new coherent wavelengths

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2,5-Dithiobiurea is used in radiopharmaceutical imaging, inhibiting enzyme function, kidney function study and to treat toxic metal poisoning. It is also used in co-crystals development and in the field of nonlinear optics to generate new coherent wavelengths

Solubility
Soluble in hot water, pyridine, triethylamine and dimethylformamide. Insoluble in cold water.

Notes
Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 142-46-1
Molecular Formula C2H6N4S2
Molecular Weight (g/mol) 150.22
MDL Number MFCD00014471
InChI Key KCOYHFNCTWXETP-UHFFFAOYSA-N
Synonym 2,5-dithiobiurea, dithiobiurea, bithiourea, dithiourea, 1,2-hydrazinedicarbothioamide, bis thiourea, hydrazodicarbothioamide, 2,5-dithiodiurea, hydrazine-1,2-bis carbothioamide, dithiocarbamoylhydrazine
PubChem CID 2724564
IUPAC Name (carbamothioylamino)thiourea
SMILES NC(=S)NNC(N)=S

Specifications

Melting Point ∼212°C (decomposition)
Quantity 25 g
Beilstein 1723178
Solubility Information Soluble in hot water,pyridine,triethylamine and dimethylformamide. Insoluble in cold water.
Formula Weight 150.22
Percent Purity 97%
Chemical Name or Material 2,5-Dithiobiurea
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 205-537-6
RTECSNumber EC1460000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only