missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dinitrophenol (wetted with ca. 20% Water) (unit weight on dry weight basis) 99.0+%, TCI America™
Supplier: TCI America D08415G
Specifications
| 2,5-Dinitrophenol (wetted with ca. 20% Water) (unit weight on dry weight basis) | |
| 106°C | |
| C6H4N2O5 | |
| 5 g | |
| UWEZBKLLMKVIPI-UHFFFAOYSA-N | |
| 2,5-dinitrophenol | |
| 9492 | |
| 184.11 | |
| Crystalline Powder |
| 329-71-5 | |
| Yellow | |
| MFCD00007108 | |
| 1320 | |
| OC1=CC(=CC=C1[N+]([O-])=O)[N+]([O-])=O | |
| 184.11 | |
| CHEBI:40810 | |
| ≥99.0% (T) |
Chemical Identifiers
| 329-71-5 | |
| 184.11 | |
| UWEZBKLLMKVIPI-UHFFFAOYSA-N | |
| CHEBI:40810 | |
| OC1=CC(=CC=C1[N+]([O-])=O)[N+]([O-])=O |
| C6H4N2O5 | |
| MFCD00007108 | |
| 9492 | |
| 2,5-dinitrophenol |
Safety and Handling
RTECSNumber : SL2900000
TSCA : No