missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylthiophene-3-boronic acid, 95%

Catalog No. p-7031458
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH32543MD 250 mg
AAH3254303 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH32543MD Supplier Thermo Scientific Chemicals Supplier No. H32543MD

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 162607-23-0 | C6H9BO2S | 156.01 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 162607-23-0
Molecular Formula C6H9BO2S
Molecular Weight (g/mol) 156.01
MDL Number MFCD09837620
InChI Key RFQHMNWNEOZRLK-UHFFFAOYSA-N
Synonym 2,5-dimethylthiophene-3-boronic acid, 2,5-dimethylthiophen-3-yl boronic acid, 2,5-dimethyl-3-thienyl boronic acid, boronic acid,b-2,5-dimethyl-3-thienyl, 2,5-dimethyl-3-thienylboronic acid, 2,5-dimethylthien-3-ylboronic acid, boronic acid, 2,5-dimethyl-3-thienyl
PubChem CID 45588123
IUPAC Name (2,5-dimethylthiophen-3-yl)boronic acid
SMILES CC1=CC(B(O)O)=C(C)S1

Specifications

Quantity 250 mg
Formula Weight 156.01
Percent Purity 95%
Chemical Name or Material 2,5-Dimethylthiophene-3-boronic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only