accessibility menu, dialog, popup

UserName

CAS RN 162607-23-0

IUPAC Name: (2,5-dimethylthiophen-3-yl)boronic acid
Synonyms: 2,5-dimethylthiophen-3-ylboronic acid
Molecular Weight (g/mol): 156.01
Molecular Formula: C6H9BO2S
InChi Key: RFQHMNWNEOZRLK-UHFFFAOYSA-N
SMILES: CC1=CC(B(O)O)=C(C)S1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results