missing translation for 'onlineSavingsMsg'
Learn More
Learn More
2,5-Dimethyltetrahydrofuran (stabilized with BHT) 98.0+%, TCI America™
Specifications
Specifications
| Color | Colorless |
| Quantity | 25 mL |
| Physical Form | Clear Liquid at 20°C |
| UN Number | 1993 |
| Boiling Point | 92°C |
| Formula Weight | 100.16 |
| Percent Purity | ≥98.0% (GC) |
| Chemical Name or Material | 2,5-Dimethyltetrahydrofuran (stabilized with BHT) |
Safety and Handling
| TSCA | No |