accessibility menu, dialog, popup

UserName

Enjoy Up to 50% Off Spotlight Deals All Year Long

Save Now

Fisher Scientific Logo
Don't have an account?  Create an Account
Products
Main Menu
Documents and Certificates

CAS RN 1003-38-9

CH3H3CO

CAS RN 1003-38-9

IUPAC Name: 2,5-dimethyloxolane
Synonyms: 2,5-dimethyltetrahydrofuran
Molecular Weight (g/mol): 100.16
Molecular Formula: C6H12O
InChi Key: OXMIDRBAFOEOQT-UHFFFAOYNA-N
SMILES: CC1CCC(C)O1
Boiling Point 
  • (1)
Percent Purity 
  • (1)
Quantity 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.