missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylterephthalic Acid 97.0+%, TCI America™

Catalog No. D22085G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D22085G 5 g
D220825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. D22085G Supplier TCI America Supplier No. D22085G

Chemical Identifiers

CAS 6051-66-7
Molecular Formula C10H10O4
Molecular Weight (g/mol) 194.186
MDL Number MFCD00020292
InChI Key FKUJGZJNDUGCFU-UHFFFAOYSA-N
PubChem CID 299989
IUPAC Name 2,5-dimethylterephthalic acid
SMILES CC1=CC(=C(C=C1C(=O)O)C)C(=O)O

Specifications

Color White
Quantity 5 g
Formula Weight 194.19
Percent Purity ≥97.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 2,5-Dimethylterephthalic Acid