missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylstyrene, 97%, stab. with 4-tert-butylcatechol

Catalog No. AAB2147603
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2147603 1 g
AAB2147606 5 g
AAB2147614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2147603 Supplier Thermo Scientific Chemicals Supplier No. B2147603
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 2039-89-6 | C10H12 | 132.206 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 2039-89-6
Molecular Formula C10H12
Molecular Weight (g/mol) 132.206
MDL Number MFCD00008614
InChI Key DBWWINQJTZYDFK-UHFFFAOYSA-N
Synonym 2,5-dimethylstyrene, styrene, 2,5-dimethyl, 1,4-dimethyl-2-vinylbenzene, 1,4-dimethyl-2-ethenylbenzene, benzene, 2-ethenyl-1,4-dimethyl, unii-rtm6p5k8ca, rtm6p5k8ca, styrene,5-dimethyl, acmc-1cjqw, 4-05-00-01385 beilstein handbook reference
PubChem CID 16265
IUPAC Name 2-ethenyl-1,4-dimethylbenzene
SMILES CC1=CC(=C(C=C1)C)C=C

Specifications

Melting Point -35°C
Density 0.904
Boiling Point 83°C to 84°C (24 mmHg)
Flash Point 63°C (145°F)
Refractive Index 1.539
Quantity 1 g
Beilstein 2203656
Formula Weight 132.21
Percent Purity 97%
Chemical Name or Material 2,5-Dimethylstyrene, stabilized with 4-tert-butylcatechol
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 218-028-9
RTECSNumber WL4460000
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only