missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrrole-3-carboxylic acid, 97%

Catalog No. p-7025246
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2063503 1 g
AAB2063506 5 g
AAB2063514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2063503 Supplier Thermo Scientific Chemicals Supplier No. B2063503

CAS: 57338-76-8 | C7H9NO2 | 139.154 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 57338-76-8
Molecular Formula C7H9NO2
Molecular Weight (g/mol) 139.154
MDL Number MFCD00203874
InChI Key VDVWTJFVFQVCFN-UHFFFAOYSA-N
Synonym 2,5-dimethylpyrrole-3-carboxylic acid, 1h-pyrrole-3-carboxylic acid, 2,5-dimethyl, 2,5-dimethyl-4-carboxypyrrole, 1h-pyrrole-3-carboxylicacid, 2,5-dimethyl
PubChem CID 292957
IUPAC Name 2,5-dimethyl-1H-pyrrole-3-carboxylic acid
SMILES CC1=CC(=C(N1)C)C(=O)O

Specifications

Melting Point 199°C to 201°C (decomposition)
Quantity 1 g
Formula Weight 139.16
Percent Purity 97%
Chemical Name or Material 2,5-Dimethylpyrrole-3-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only