accessibility menu, dialog, popup

UserName

CH3NHCH3OO

CAS RN 57338-76-8

IUPAC Name: 2,5-dimethyl-1H-pyrrole-3-carboxylate
Synonyms: 2,5-dimethyl-1H-pyrrole-3-carboxylate
Molecular Weight (g/mol): 138.15
Molecular Formula: C7H8NO2
InChi Key: VDVWTJFVFQVCFN-UHFFFAOYSA-M
SMILES: CC1=CC(C([O-])=O)=C(C)N1
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results