missing translation for 'onlineSavingsMsg'
Learn More

2,5-Dimethylpyrazine 98.0+%, TCI America™

Catalog No. D217125ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
D217125ML 25 mL
D2171100ML 100 mL
D2171500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. D217125ML Supplier TCI America Supplier No. D217125ML

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 123-32-0
Molecular Formula C6H8N2
Molecular Weight (g/mol) 108.144
MDL Number MFCD00006147
InChI Key LCZUOKDVTBMCMX-UHFFFAOYSA-N
Synonym 2,5-dimethyl pyrazine, pyrazine, 2,5-dimethyl, 2,5-dimethylpiazine, 2,5-dimethyl-1,4-diazine, 2,5-dimethylparadiazine, 2,5-dimethyl-pyrazine, unii-v99y0muy1q, fema no. 3272, ccris 2929, natural
PubChem CID 31252
IUPAC Name 2,5-dimethylpyrazine
SMILES CC1=CN=C(C=N1)C

Specifications

Color Yellow
Boiling Point 155°C
Quantity 25 mL
Formula Weight 108.14
Percent Purity ≥98.0% (GC,T)
Physical Form Liquid
Chemical Name or Material 2,5-Dimethylpyrazine
EINECSNumber (5)-0950
RTECSNumber UQ2800000
TSCA Yes